Learn More
Medchemexpress LLC 9-ethyl-9H-carbazol-3-amine | 132-32-1 | MFCD00004964 | 99.6% | 210.28 g/mol | C14H14N2 | 100 G
SDP
Catalog No. 5000302400
Encompass_Preferred
3-Amino-9-ethylcarbazole (9-ethyl-9H-carbazol-3-amine) is an organic small molecule used as a chromogenic peroxidase substrate in immunohistochemistry and as a building block in organic electronic materials and synthetic chemistry. It is typically supplied as a solid powder with molecular formula C14H14N2 and molecular weight 210.28 g/mol.
  • Chromogenic peroxidase substrate for histochemical and blotting assays.
  • Versatile building block for organic electronics and small-molecule synthesis.
  • High purity suitable for analytical and synthetic use.
  • Available in multiple pack sizes to support research-scale needs.
  • Certificate of analysis available to document batch quality.
Catalog No. 50-003-02400 Supplier Medchemexpress LLC Supplier No. HYW014386100G
3-Amino-9-ethylcarbazole (9-ethyl-9H-carbazol-3-amine) is an organic small molecule used as a chromogenic peroxidase substrate in immunohistochemistry and as a building block in organic electronic materials and synthetic chemistry. It is typically supplied as a solid powder with molecular formula C14H14N2 and molecular weight 210.28 g/mol.
  • Chromogenic peroxidase substrate for histochemical and blotting assays.
  • Versatile building block for organic electronics and small-molecule synthesis.
  • High purity suitable for analytical and synthetic use.
  • Available in multiple pack sizes to support research-scale needs.
  • Certificate of analysis available to document batch quality.