Learn More
Medchemexpress LLC HY-119413 100mg Medchemexpress, 9-Ethyladenine CAS:2715-68-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY119413100MG
Encompass_Preferred
Medchemexpress, HY-119413 100mg 9-Ethyladenine CAS:2715-68-6 9-Ethyladenine is a partially effective inhibitor of APRT (adenine phosphoribosyltransferase). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9534