Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Ethylcarbazole-3-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. E11535G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1153-5G 5 g
E11531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E1153-5G Supplier TCI America Supplier No. E11535G
Add to Cart
Add to Cart

Specifications

CAS 669072-93-9
Color White
Molecular Formula C14H14BNO2
Quantity 5 g
MDL Number MFCD09743121
Synonym 9-ethyl-9 h-carbazol-3-yl boronic acid, 9-ethyl-3-carbazole boronic acid, boronic acid, 9-ethyl-9 h-carbazol-3-yl, ksc924s3d, 9-ethylcarbazole-3-boronic acid
InChI Key JUBKLYCRFZTXBA-UHFFFAOYSA-N
SMILES B(C1=CC2=C(C=C1)N(C3=CC=CC=C32)CC)(O)O
IUPAC Name (9-ethylcarbazol-3-yl)boronic acid
Molecular Weight (g/mol) 239.081
PubChem CID 45382262
Formula Weight 239.08
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.