Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Ethylcarbazole, 99%

Catalog No. AAA116530B
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA116530B 1000 g
AAA1165322 100 g
AAA1165330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA116530B Supplier Thermo Scientific Chemicals Supplier No. A116530B
Only null left
Add to Cart

CAS: 86-28-2 | C14H13N | 195.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-28-2
Molecular Formula C14H13N
Molecular Weight (g/mol) 195.27
MDL Number MFCD00004967
InChI Key PLAZXGNBGZYJSA-UHFFFAOYSA-N
Synonym n-ethylcarbazole, 9-ethyl-9h-carbazole, 9h-carbazole, 9-ethyl, carbazole, 9-ethyl, n-ethylcarbazol, n-ethyl carbazole, unii-6ak165l0ro, ccris 6847, 9h-ethylcarbazole, 9-ethyl carbazole
PubChem CID 6836
IUPAC Name 9-ethylcarbazole
SMILES CCN1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 67°C to 71°C
Boiling Point 166°C (4 mmHg)
Flash Point 186°C (367°F)
Quantity 1000 g
Beilstein 148115
Formula Weight 195.27
Percent Purity 99%
Chemical Name or Material 9-Ethylcarbazole
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 201-660-4
RTECSNumber FE6225700
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.