Learn More
Medchemexpress LLC 9-ethylcarbazole (N-ethylcarbazole) | 86-28-2 | MFCD00004967 | 100.0% | 195.26 g/mol | C14H13N | 100 G
SDP
Catalog No. 5000302417
Encompass_Preferred
9-Ethylcarbazole (N-ethylcarbazole) is an aromatic nitrogen-containing heterocycle used in materials and energy research. It is commonly studied as a liquid organic hydrogen carrier and used as a reagent or building block in organic synthesis, supplied as a high-purity laboratory chemical suitable for analytical and synthetic workflows.
  • Used as a liquid organic hydrogen carrier for hydrogen storage research.
  • High purity reagent appropriate for analytical and synthetic applications.
  • Available in laboratory-scale package sizes including 100 g.
  • Stable aromatic heterocycle; chemical formula C14H13N; molar mass 195.26 g/mol.
  • Suitable for materials science and organic chemistry research.
Catalog No. 50-003-02417 Supplier Medchemexpress LLC Supplier No. HYW015003100G
9-Ethylcarbazole (N-ethylcarbazole) is an aromatic nitrogen-containing heterocycle used in materials and energy research. It is commonly studied as a liquid organic hydrogen carrier and used as a reagent or building block in organic synthesis, supplied as a high-purity laboratory chemical suitable for analytical and synthetic workflows.
  • Used as a liquid organic hydrogen carrier for hydrogen storage research.
  • High purity reagent appropriate for analytical and synthetic applications.
  • Available in laboratory-scale package sizes including 100 g.
  • Stable aromatic heterocycle; chemical formula C14H13N; molar mass 195.26 g/mol.
  • Suitable for materials science and organic chemistry research.