Learn More
Medchemexpress LLC 9-ethylcarbazole (N-ethylcarbazole) | 86-28-2 | MFCD00004967 | 100.0% | 195.26 g/mol | C14H13N | 25 G
SDP
Catalog No. 5000303593
Encompass_Preferred
9-Ethylcarbazole (N-ethylcarbazole) is an organic heterocyclic compound used as a hydrogen-storage material and research reagent. Provided as a high-purity solid for laboratory use, it is employed in materials research, dehydrogenation studies, and synthetic chemistry. Key identifiers include molecular formula C14H13N, molecular weight 195.26 g/mol, and CAS 86-28-2.
  • High purity suitable for research applications.
  • Solid, easy-to-handle material for lab workflows.
  • Used in hydrogen storage and dehydrogenation studies.
  • Applicable to catalysis research and organic synthesis.
  • Compatible with common organic solvents.
  • Available in multiple pack sizes to suit experimental needs.
Catalog No. 50-003-03593 Supplier Medchemexpress LLC Supplier No. 25GHYW015003
9-Ethylcarbazole (N-ethylcarbazole) is an organic heterocyclic compound used as a hydrogen-storage material and research reagent. Provided as a high-purity solid for laboratory use, it is employed in materials research, dehydrogenation studies, and synthetic chemistry. Key identifiers include molecular formula C14H13N, molecular weight 195.26 g/mol, and CAS 86-28-2.
  • High purity suitable for research applications.
  • Solid, easy-to-handle material for lab workflows.
  • Used in hydrogen storage and dehydrogenation studies.
  • Applicable to catalysis research and organic synthesis.
  • Compatible with common organic solvents.
  • Available in multiple pack sizes to suit experimental needs.