Learn More
Cayman Chemical 9-Ethylguanine, Purity: ≥98%, CAS: 879-08-3, 100 mg
SDP
Catalog No. 502965033
Encompass_Preferred
A model nucleobase that is used to study DNA interactions with organometallic complexes, especially those designed to target tumors
Catalog No. 50-296-5033 Supplier Cayman Chemical Supplier No. 18738100 MG
A model nucleobase that is used to study DNA interactions with organometallic complexes, especially those designed to target tumors