Learn More
Cayman Chemical 9-Ethylguanine, Purity: ≥98%, CAS: 879-08-3, 50 mg
SDP
Catalog No. 502967799
Encompass_Preferred
A model nucleobase that is used to study DNA interactions with organometallic complexes, especially those designed to target tumors
Catalog No. 50-296-7799 Supplier Cayman Chemical Supplier No. 1873850 MG
A model nucleobase that is used to study DNA interactions with organometallic complexes, especially those designed to target tumors