Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Fluorenone-1-carbonyl chloride, 95%, Thermo Scientific™

Catalog No. AAH3690303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3690303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3690303 Supplier Thermo Scientific Chemicals Supplier No. H3690303

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55341-62-3
Molecular Formula C14H7ClO2
Molecular Weight (g/mol) 242.66
MDL Number MFCD20485912
InChI Key ZNXLQKIGKKVHNF-UHFFFAOYSA-N
Synonym 9-oxo-9h-fluorene-1-carbonyl chloride, 3-trifluoromethylsulfanyl-benzoyl chloride, fluorene-9-one-1-carboxylic acid chloride
PubChem CID 12209857
IUPAC Name 9-oxofluorene-1-carbonyl chloride
SMILES ClC(=O)C1=CC=CC2=C1C(=O)C1=CC=CC=C21

Specifications

Melting Point 126°C to 128°C
Color Orange
Quantity 1 g
UN Number UN3261
Sensitivity Moisture Sensitive
Formula Weight 242.66
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 9-Fluorenone-1-carbonyl chloride
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.