Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Fluorenone-1-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. F05905G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05905G 5 g
F05901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05905G Supplier TCI America Supplier No. F05905G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1573-92-8
Molecular Formula C14H8O3
Molecular Weight (g/mol) 224.22
MDL Number MFCD00011537
InChI Key CBEFMGJHEKAMNI-UHFFFAOYSA-N
Synonym 9-oxo-9h-fluorene-1-carboxylic acid, 9-fluorenone-1-carboxylic acid, 9-oxo-9h-fluorenecarboxylic acid, 9-oxofluorenecarboxylic acid, 9-fluorenone-1-carboxylicacid, 9h-fluorene-1-carboxylic acid, 9-oxo, maybridge1_002192, 9-oxo-1-fluorenecarboxylic acid, 9-oxo-9h-fluorene-1-carboxylicacid, 9h-fluorenecarboxylic acid, 9-oxo
PubChem CID 74083
IUPAC Name 9-oxo-9H-fluorene-1-carboxylic acid
SMILES OC(=O)C1=CC=CC2=C1C(=O)C1=CC=CC=C21

Specifications

Melting Point 198°C
Quantity 5 g
Formula Weight 224.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9-Fluorenone-1-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.