Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Fluorenone-2-carboxylic Acid 96.0+%, TCI America™
SDP

Catalog No. F026310G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F026310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F026310G Supplier TCI America Supplier No. F026310G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 784-50-9
Molecular Formula C14H8O3
Molecular Weight (g/mol) 224.22
MDL Number MFCD00001156
InChI Key BJCTXUUKONLPPK-UHFFFAOYSA-N
Synonym 9-fluorenone-2-carboxylic acid, 9-oxo-9h-fluorene-2-carboxylic acid, fluorenone-2-carboxylic acid, 9-oxofluoren-2-carboxylic acid, 9h-fluorene-2-carboxylic acid, 9-oxo, 9-fluorenone-2-carboxylicacid, 9-oxo-2-fluorenecarboxylic acid, bjctxuukonlppk-uhfffaoysa, 9-oxo-2-fluorene carboxylic acid, 9-oxo-fluorene-2-carboxylic acid
PubChem CID 69913
IUPAC Name 9-oxo-9H-fluorene-2-carboxylic acid
SMILES OC(=O)C1=CC=C2C3=CC=CC=C3C(=O)C2=C1

Specifications

Quantity 10 g
Formula Weight 224.22
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9-Fluorenone-2-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.