Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Fluorenone, 98+%

Catalog No. aaa1291022
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1291022 100 g
AAA1291036 500 g
AAA129100E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1291022 Supplier Thermo Scientific Chemicals Supplier No. A1291022
Only null left
Add to Cart

CAS: 486-25-9 | C13H8O | 180.206 g/mol

9-Fluorenone is used in the preparation of antimalarial drugs. It is a fluorene derivative. Further, it is used in functional polymer and in dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Fluorenone is used in the preparation of antimalarial drugs. It is a fluorene derivative. Further, it is used in functional polymer and in dyes.

Solubility
Soluble in water, alcohol, acetone, ether and benzene.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 486-25-9
Molecular Formula C13H8O
Molecular Weight (g/mol) 180.206
MDL Number MFCD00001141
InChI Key YLQWCDOCJODRMT-UHFFFAOYSA-N
Synonym 9-fluorenone, 9h-fluoren-9-one, fluorenone, 9-oxofluorene, diphenylene ketone, unii-az9t83s2aq, ccris 593, 9h-fluorene-9-one, az9t83s2aq, 9-fluoreneone
PubChem CID 10241
ChEBI CHEBI:17922
IUPAC Name fluoren-9-one
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2=O

Specifications

Melting Point 82°C to 85°C
Density 0.9
Boiling Point 342°C
Flash Point 163°C (325°F)
Assay Percent Range 98%
Quantity 100 g
Beilstein 1636531
Solubility Information Soluble in water,alcohol,acetone,ether and benzene.
Formula Weight 180.21
Percent Purity ≥98%
Chemical Name or Material 9-Fluorenone
Show More Show Less
EINECSNumber 207-630-7
RTECSNumber LL8925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.