Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Fluorenone, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass Bottle
Plastic bottle
486-25-9
C13H8O
180.21
YLQWCDOCJODRMT-UHFFFAOYSA-N
9-fluorenone,9h-fluoren-9-one,fluorenone,9-oxofluorene,diphenylene ketone,unii-az9t83s2aq,ccris 593,9h-fluorene-9-one,az9t83s2aq,9-fluoreneone
10241
CHEBI:17922
fluoren-9-one
C1=CC=C2C(=C1)C3=CC=CC=C3C2=O
486-25-9
C13H8O
180.21
YLQWCDOCJODRMT-UHFFFAOYSA-N
9-fluorenone,9h-fluoren-9-one,fluorenone,9-oxofluorene,diphenylene ketone,unii-az9t83s2aq,ccris 593,9h-fluorene-9-one,az9t83s2aq,9-fluoreneone
10241
CHEBI:17922
fluoren-9-one
C1=CC=C2C(=C1)C3=CC=CC=C3C2=O
Quantity 100g
Flash Point 163°C
Melting Point 81.0°C to 85.0°C
Packaging Plastic bottle
Color Yellow
Boiling Point 342.0°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Beilstein 07,465
Solubility Information Solubility in water: insoluble.
Formula Weight 180.21
Physical Form Crystalline Powder or Flakes
Percent Purity ≥99%
Chemical Name or Material 9-Fluorenone
Show More Show Less