Learn More
Medchemexpress LLC 9-Fluorenylmethanol | 24324-17-2 | 99.0% | 196.25 | 500 G
SDP
Catalog No. 5000366419
Encompass_Preferred
9-Fluorenylmethanol is an N-protecting reagent primarily used in peptide synthesis. This product is suitable for laboratory centrifugation and storage.
  • N-protecting reagent
  • Used in peptide synthesis
  • Solid appearance
  • Off-white to light yellow color
Catalog No. 50-003-66419 Supplier Medchemexpress LLC Supplier No. HYW009876500G
9-Fluorenylmethanol is an N-protecting reagent primarily used in peptide synthesis. This product is suitable for laboratory centrifugation and storage.
  • N-protecting reagent
  • Used in peptide synthesis
  • Solid appearance
  • Off-white to light yellow color