Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9-fluorenylmethoxycarbonyl-4-tert-butyl-L-phenylalanine | 213383-02-9 | 99.9% | 443.53 g/mol | C28H29NO4 | 1 G
SDP

Catalog No. 5000209772
Encompass_Preferred

Fmoc-Phe(4-tBu)-OH is an Fmoc-protected 4-tert-butyl phenylalanine derivative used as a building block in peptide synthesis. Supplied as a white to off-white solid with high reported purity, it is intended for research use only.

  • Molecular formula: C28H29NO4.
  • Molecular weight: 443.53 g/mol.
  • CAS number: 213383-02-9.
  • Purity: 99.9%.
  • Appearance: white to off-white solid.
  • Storage: store at room temperature (solid); in solution, store at -80°C for up to 2 years or -20°C for up to 1 year.
  • Typical quantity: 1 G.

Catalog No. 50-002-09772 Supplier Medchemexpress LLC Supplier No. HYW0487301G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Phe(4-tBu)-OH is an Fmoc-protected 4-tert-butyl phenylalanine derivative used as a building block in peptide synthesis. Supplied as a white to off-white solid with high reported purity, it is intended for research use only.

  • Molecular formula: C28H29NO4.
  • Molecular weight: 443.53 g/mol.
  • CAS number: 213383-02-9.
  • Purity: 99.9%.
  • Appearance: white to off-white solid.
  • Storage: store at room temperature (solid); in solution, store at -80°C for up to 2 years or -20°C for up to 1 year.
  • Typical quantity: 1 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.