Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9-Fluorenylmethoxycarbonyl 4-(tert-butyl)phenylalanine | 213383-02-9 | MFCD02094466 | 99.9% | 443.53 g/mol | C28H29NO4 | 25 G
SDP

Catalog No. 5000209504
Encompass_Preferred

Fmoc-Phe(4-tBu)-OH is an Fmoc-protected derivative of 4-tert-butyl phenylalanine used as a building block in solid-phase and solution-phase peptide synthesis. It offers high chemical purity and defined molecular properties suitable for incorporation into synthetic peptides.

  • Fmoc-protected amino acid for peptide synthesis.
  • High purity: 99.9%.
  • Molecular weight: 443.53 g/mol.
  • Molecular formula: C28H29NO4.
  • Available in multiple package sizes, including 25 G.

Catalog No. 50-002-09504 Supplier Medchemexpress LLC Supplier No. 25GHYW048730
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Phe(4-tBu)-OH is an Fmoc-protected derivative of 4-tert-butyl phenylalanine used as a building block in solid-phase and solution-phase peptide synthesis. It offers high chemical purity and defined molecular properties suitable for incorporation into synthetic peptides.

  • Fmoc-protected amino acid for peptide synthesis.
  • High purity: 99.9%.
  • Molecular weight: 443.53 g/mol.
  • Molecular formula: C28H29NO4.
  • Available in multiple package sizes, including 25 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.