Learn More
Medchemexpress LLC 9-Fluorenylmethoxycarbonyl glutamic acid methyl ester | 145038-49-9 | MFCD22200617 | 99.7% | 383.40 g/mol | C21H21NO6 | 10 G
SDP
Catalog No. 5000208492
Encompass_Preferred
Fmoc-Glu-OMe is the Fmoc-protected methyl ester derivative of glutamic acid, used as a research reagent. It has been reported to show antibacterial activity and to form gels in AgNO3 solution, and has been described as a mouldable wound-healing biomaterial.
  • Fmoc-protected amino acid derivative.
  • Shows antibacterial activity in AgNO3 solution.
  • Exhibits gelation enabling mouldable biomaterial formation.
  • Supplied as a powder for laboratory research use.
  • Available in a 10 G pack size; high-purity grade suitable for research.
Catalog No. 50-002-08492 Supplier Medchemexpress LLC Supplier No. HYW04198810G
Fmoc-Glu-OMe is the Fmoc-protected methyl ester derivative of glutamic acid, used as a research reagent. It has been reported to show antibacterial activity and to form gels in AgNO3 solution, and has been described as a mouldable wound-healing biomaterial.
  • Fmoc-protected amino acid derivative.
  • Shows antibacterial activity in AgNO3 solution.
  • Exhibits gelation enabling mouldable biomaterial formation.
  • Supplied as a powder for laboratory research use.
  • Available in a 10 G pack size; high-purity grade suitable for research.