Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9-fluorenylmethyl carbamate | 84418-43-9 | MFCD00237376 | 99.8% | 239.27 g·mol⁻¹ | C15H13NO2 | 25 G
SDP

Catalog No. 5000303090
Encompass_Preferred

9-Fluorenylmethyl carbamate (Fmoc-NH2) is an Fmoc-protected amide reagent supplied as a white to off-white solid. It is used as a building block in organic and peptide synthesis and has reported use as a photobase initiator for preparing organosilane-based proton exchange membranes. The product is supplied in laboratory pack sizes and is accompanied by analytical documentation.

  • High purity (~99.8%).
  • Molecular formula C15H13NO2.
  • Molecular weight 239.27 g·mol⁻¹.
  • Appearance white to off-white solid.
  • Storage: room temperature for solid; in solution -20°C for 1 year or -80°C for 2 years.
  • Available documents: data sheet, certificate of analysis, SDS, H NMR, RP-HPLC.
  • Typical applications include organic synthesis, peptide chemistry, and photobase initiation.

Catalog No. 50-003-03090 Supplier Medchemexpress LLC Supplier No. 25GHYW015987
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

9-Fluorenylmethyl carbamate (Fmoc-NH2) is an Fmoc-protected amide reagent supplied as a white to off-white solid. It is used as a building block in organic and peptide synthesis and has reported use as a photobase initiator for preparing organosilane-based proton exchange membranes. The product is supplied in laboratory pack sizes and is accompanied by analytical documentation.

  • High purity (~99.8%).
  • Molecular formula C15H13NO2.
  • Molecular weight 239.27 g·mol⁻¹.
  • Appearance white to off-white solid.
  • Storage: room temperature for solid; in solution -20°C for 1 year or -80°C for 2 years.
  • Available documents: data sheet, certificate of analysis, SDS, H NMR, RP-HPLC.
  • Typical applications include organic synthesis, peptide chemistry, and photobase initiation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.