Learn More
Medchemexpress LLC 9-fluorenylmethyl carbamate | 84418-43-9 | MFCD00237376 | 99.9% | 239.27 g/mol | C15H13NO2 | 100 G
SDP
Catalog No. 5000303901
Encompass_Preferred
9-Fluorenylmethyl carbamate (Fmoc-NH2) is an Fmoc-protected amide supplied as a solid reagent for laboratory synthesis and materials research. It functions as an Fmoc-protected amine and can act as a photobase initiator in the preparation of organosilane-based proton exchange membranes and related polymer applications.
  • High chemical purity suitable for research use.
  • Fmoc-protected amine functionality for orthogonal deprotection strategies.
  • Used as a photobase initiator in organosilane and polymer chemistry.
  • Stable solid form for ease of handling and storage.
  • Molecular formula C15H13NO2, molecular weight 239.27 g/mol.
Catalog No. 50-003-03901 Supplier Medchemexpress LLC Supplier No. HYW015987100G
9-Fluorenylmethyl carbamate (Fmoc-NH2) is an Fmoc-protected amide supplied as a solid reagent for laboratory synthesis and materials research. It functions as an Fmoc-protected amine and can act as a photobase initiator in the preparation of organosilane-based proton exchange membranes and related polymer applications.
  • High chemical purity suitable for research use.
  • Fmoc-protected amine functionality for orthogonal deprotection strategies.
  • Used as a photobase initiator in organosilane and polymer chemistry.
  • Stable solid form for ease of handling and storage.
  • Molecular formula C15H13NO2, molecular weight 239.27 g/mol.