Learn More
Medchemexpress LLC 9-fluorenylmethyl carbamate (Fmoc-NH2) | 84418-43-9 | MFCD00270532 | 99.9% | 239.27 g/mol | C15H13NO2 | 50 G
SDP
Catalog No. 5000300278
Encompass_Preferred
9-Fluorenylmethyl carbamate (Fmoc-NH2) is an Fmoc-protected primary amine used as a reagent in organic and peptide synthesis and as a photobase initiator in materials research. Supplied as a solid with high listed purity, it serves as a building block for introducing Fmoc protection or for preparing photobase-active organosilane materials.
  • Fmoc-protected primary amine for orthogonal protection in synthesis.
  • Functions as a photobase initiator for organosilane-based proton exchange membranes.
  • High listed purity suitable for research applications.
  • Available in multiple pack sizes for scale flexibility.
  • Molecular weight 239.27 g/mol and chemical formula C15H13NO2.
Catalog No. 50-003-00278 Supplier Medchemexpress LLC Supplier No. HYW01598750G
9-Fluorenylmethyl carbamate (Fmoc-NH2) is an Fmoc-protected primary amine used as a reagent in organic and peptide synthesis and as a photobase initiator in materials research. Supplied as a solid with high listed purity, it serves as a building block for introducing Fmoc protection or for preparing photobase-active organosilane materials.
  • Fmoc-protected primary amine for orthogonal protection in synthesis.
  • Functions as a photobase initiator for organosilane-based proton exchange membranes.
  • High listed purity suitable for research applications.
  • Available in multiple pack sizes for scale flexibility.
  • Molecular weight 239.27 g/mol and chemical formula C15H13NO2.