Learn More
Apexbio Technology LLC 9-Fluorenylmethyl carbazate 35661-51-9 250mg
SDP
Catalog No. 5000092851
Encompass_Preferred
9-Fluorenylmethyl carbazate (CAS 35661-51-9) is a fluorescent labeling reagent used for carbohydrate detection and glycan structural characterization It facilitates fluorescence-based analytical methods by derivatizing glycans enabling their quantitative and qualitative analysis through chromatographic or electrophoretic techniques coupled with fluorescence detection Based on these properties 9-fluorenylmethyl carbazate holds research potential in the analysis of glycoproteins and polysaccharides supporting detailed evaluation of glycosylation patterns in biomedical research
Catalog No. 50-000-92851 Supplier Apexbio Technology LLC Supplier No. A4779250
9-Fluorenylmethyl carbazate (CAS 35661-51-9) is a fluorescent labeling reagent used for carbohydrate detection and glycan structural characterization It facilitates fluorescence-based analytical methods by derivatizing glycans enabling their quantitative and qualitative analysis through chromatographic or electrophoretic techniques coupled with fluorescence detection Based on these properties 9-fluorenylmethyl carbazate holds research potential in the analysis of glycoproteins and polysaccharides supporting detailed evaluation of glycosylation patterns in biomedical research