Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9-Fluorenylmethyl carbazate | 35661-51-9 | MFCD00239419 | 99.0% | 254.28 g/mol | C15H14N2O2 | 5 G
SDP

Catalog No. 5000299678
Encompass_Preferred

9-Fluorenylmethyl carbazate is a fluorescent labeling reagent used to derivatize glycans and other carbonyl-containing compounds for sensitive fluorimetric detection. It is supplied as a white to off-white solid and as a 10 mM solution in DMSO, and is intended for research use only.

  • Fluorophore reagent for fluorimetric detection of glycans.
  • Available as solid and as a 10 mM solution in DMSO.
  • High reported purity suitable for analytical derivatization.
  • Molecular formula C15H14N2O2; molecular weight 254.28 g/mol.
  • Batch-specific purity information provided.

Catalog No. 50-002-99678 Supplier Medchemexpress LLC Supplier No. HYW0178735G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

9-Fluorenylmethyl carbazate is a fluorescent labeling reagent used to derivatize glycans and other carbonyl-containing compounds for sensitive fluorimetric detection. It is supplied as a white to off-white solid and as a 10 mM solution in DMSO, and is intended for research use only.

  • Fluorophore reagent for fluorimetric detection of glycans.
  • Available as solid and as a 10 mM solution in DMSO.
  • High reported purity suitable for analytical derivatization.
  • Molecular formula C15H14N2O2; molecular weight 254.28 g/mol.
  • Batch-specific purity information provided.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.