Learn More
Medchemexpress LLC 9-fluorenylmethyl carbazate | 35661-51-9 | MFCD00239419 | 99.9% | 254.28 g/mol | C15H14N2O2 | 10 G
SDP
Catalog No. 5000301406
Encompass_Preferred
9-Fluorenylmethyl carbazate is a fluorescent labeling reagent used for derivatization and fluorimetric detection of glycans and other analytes. It is supplied for research use and is suitable for HPLC derivatization and fluorescence-based assays.
  • Fluorimetric reagent for glycan detection and HPLC derivatization.
  • High purity: 99.9%.
  • Chemical formula C15H14N2O2; molecular weight 254.28 g/mol.
  • Available in multiple pack sizes, including 10 g.
  • Stable under recommended refrigerated or frozen storage conditions.
  • Intended for research use only.
Catalog No. 50-003-01406 Supplier Medchemexpress LLC Supplier No. HYW01787310G
9-Fluorenylmethyl carbazate is a fluorescent labeling reagent used for derivatization and fluorimetric detection of glycans and other analytes. It is supplied for research use and is suitable for HPLC derivatization and fluorescence-based assays.
  • Fluorimetric reagent for glycan detection and HPLC derivatization.
  • High purity: 99.9%.
  • Chemical formula C15H14N2O2; molecular weight 254.28 g/mol.
  • Available in multiple pack sizes, including 10 g.
  • Stable under recommended refrigerated or frozen storage conditions.
  • Intended for research use only.