Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Fluorenylmethyl chloroformate, 98%

Catalog No. AC170940010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170940010 1 g Glass bottle
AC170940050 5 g Glass bottle
AC170940250 25 g Glass bottle
AC170941000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC170940010 Supplier Thermo Scientific Chemicals Supplier No. 170940010
  • $45.25
    $38.65 / Each of 1

    Save $6.60

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 28920-43-6 | C15H11ClO2 | 258.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28920-43-6
Molecular Formula C15H11ClO2
Molecular Weight (g/mol) 258.69
MDL Number MFCD00001138
InChI Key IRXSLJNXXZKURP-UHFFFAOYSA-N
Synonym 9-fluorenylmethyl chloroformate, fmoc-cl, fmoc-chloride, fmoc chloride, 9h-fluoren-9-ylmethyl chloroformate, carbonochloridic acid, 9h-fluoren-9-ylmethyl ester, 9-fluorenylmethoxycarbonyl chloride, 9-fluorenylmethylchloroformate, 9h-fluoren-9-yl methyl carbonochloridate, 9h-fluoren-9-ylmethoxy carbonyl chloride
PubChem CID 34367
IUPAC Name 9H-fluoren-9-ylmethyl carbonochloridate
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 63.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 4190
Solubility Information Solubility in water: decomposes in water.,acetone
Formula Weight 258.69
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 9-Fluorenylmethyl chloroformate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
EINECSNumber 249-313-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.