Learn More
Medchemexpress LLC 9-fluoro-11β,17,21-trihydroxypregna-1,4-diene-3,20-dione 21-acetate | 338-98-7 | MFCD00468096 | 98.6% | C23H29FO6 | 10MG
SDP
Catalog No. 5000260385
Encompass_Preferred
Isoflupredone acetate is a synthetic corticosteroid acetate used as an analytical reference standard for research and analytical applications such as HPLC, GC, and MS. Supplied as a high-purity solid, it is intended for assay development, calibration, and method validation in pharmaceutical and veterinary research.
  • Analytical standard for HPLC, GC, and MS
  • High reported purity (~98.5%)
  • Suitable for assay development and method validation
  • Available in small milligram pack sizes for reference use
  • Provided with batch-specific certificate of analysis
Catalog No. 50-002-60385 Supplier Medchemexpress LLC Supplier No. HY13411610MG
Isoflupredone acetate is a synthetic corticosteroid acetate used as an analytical reference standard for research and analytical applications such as HPLC, GC, and MS. Supplied as a high-purity solid, it is intended for assay development, calibration, and method validation in pharmaceutical and veterinary research.
  • Analytical standard for HPLC, GC, and MS
  • High reported purity (~98.5%)
  • Suitable for assay development and method validation
  • Available in small milligram pack sizes for reference use
  • Provided with batch-specific certificate of analysis