Learn More
Medchemexpress LLC 9-Heptadecanone 25g
SDP
Catalog No. 5000204635
Encompass_Preferred
9-Heptadecanone (Heptadecan-9-one) is an aliphatic molecule with a central ketone on a C17 chain The ketone may react with amines in reductive aminations to link this molecule to larger lipid structures
Catalog No. 50-002-04635 Supplier Medchemexpress LLC Supplier No. 25GHYW099634
9-Heptadecanone (Heptadecan-9-one) is an aliphatic molecule with a central ketone on a C17 chain The ketone may react with amines in reductive aminations to link this molecule to larger lipid structures