Learn More
Medchemexpress LLC 9-Hexadecenoic acid, methyl ester, (9Z)- | 1120-25-8 | 98.1% | 268.43 | 250 MG
SDP
Catalog No. 5000453285
Encompass_Preferred
Methyl palmitoleate, a fatty acid methyl ester, is an analogue of Palmitoleate with cytoprotective and growth-promoting properties. It inhibits the formation and aggregation of wax crystals and improves the low-temperature fluidity of biodiesel as a biodiesel additive.
  • Fatty acid methyl ester
  • Analogue of Palmitoleate
  • Cytoprotective properties
  • Growth-promoting properties
  • Inhibits the formation and aggregation of wax crystals
  • Improves the low-temperature fluidity of biodiesel
Catalog No. 50-004-53285 Supplier Medchemexpress LLC Supplier No. 250MGHYW011688
Methyl palmitoleate, a fatty acid methyl ester, is an analogue of Palmitoleate with cytoprotective and growth-promoting properties. It inhibits the formation and aggregation of wax crystals and improves the low-temperature fluidity of biodiesel as a biodiesel additive.
  • Fatty acid methyl ester
  • Analogue of Palmitoleate
  • Cytoprotective properties
  • Growth-promoting properties
  • Inhibits the formation and aggregation of wax crystals
  • Improves the low-temperature fluidity of biodiesel