Learn More
AdipoGen 9-HMe-10-carbamoylacridan
SDP
Supplier:  AdipoGen CDXH0118M025
Encompass_Preferred
Chemical. CAS 68011-71-2. Formula C15H14N2O2. MW 254.28. Synthetic Major intermediate in carbamazepine metabolism. This carbamazepine metabolite can be used as a reference compound.
Catalog No. 50-256-0714