Learn More
Cayman Chemical 9-hydroxy Stearic Acid, Purity: ≥95%, CAS: 3384-24-5, 10 mg
SDP
Catalog No. 502968168
Encompass_Preferred
A hydroxy fatty acid and an active metabolite of 9-PAHSA; inhibits HDAC1 in HT-29 cell lysates at 5 µM; inhibits proliferation of, and induces cell cycle arrest at the G0/G1 phase in, HT-29 cells at 100 µM
Catalog No. 50-296-8168 Supplier Cayman Chemical Supplier No. 1801610 MG
A hydroxy fatty acid and an active metabolite of 9-PAHSA; inhibits HDAC1 in HT-29 cell lysates at 5 µM; inhibits proliferation of, and induces cell cycle arrest at the G0/G1 phase in, HT-29 cells at 100 µM