Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 9-hydroxynonanoic acid

Catalog No. CDS018651 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS018651 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS018651 Supplier Sigma Aldrich Supplier No. CDS01865110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N,n-dimethylhexadecylamine is a swelling agent useful for the enlargement of the size of mesopores of colloidal silica nanoparticles.n,n-dimethylhexadecylamine may be employed in the following studies:
  • As pore size mediator for the synthesis of monodispersed mesoporous silica nanoparticles
  • As swelling agent in the fabrication of colloidal mesoporous silica nanoparticles of a tunable mesopore size
  • Preparation of 1,4-bis(n-hexadecyl-n,n-dimethylammonium)butane dibromide, gemini surfactant

Specifications

Molecular Formula C9H18O3
Linear Formula C9H18O3
MDL Number MFCD00792445
Quantity 100 mL
Molecular Weight (g/mol) 174.24
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.