Learn More
Medchemexpress LLC 9-Hydroxyphenanthrene | 484-17-3 | 98.6% | 194.23 | 250 MG
SDP
Catalog No. 5000347593
Encompass_Preferred
9-Phenanthrol (9-Hydroxyphenanthrene) is a selective TRPM4 inhibitor with an IC50 of 17 μM. It demonstrates no inhibitory activity on TRPM5, TRPC6, and CFTR. This compound can be utilized for research into ischemia-reperfusion injury, with studies showing it can reduce myocardial dysfunction in isolated rat hearts subjected to ischemia and reperfusion.
  • Selective TRPM4 inhibitor with an IC50 of 17 μM.
  • Shows no inhibitory activity on TRPM5, TRPC6, and CFTR.
  • Useful for research into ischemia-reperfusion injury.
  • Can reduce myocardial dysfunction in isolated rat hearts.
  • Leads to significant improvement in heart function.
  • Reduces infarct size in affected hearts.
Catalog No. 50-003-47593 Supplier Medchemexpress LLC Supplier No. 250MGHY108457
9-Phenanthrol (9-Hydroxyphenanthrene) is a selective TRPM4 inhibitor with an IC50 of 17 μM. It demonstrates no inhibitory activity on TRPM5, TRPC6, and CFTR. This compound can be utilized for research into ischemia-reperfusion injury, with studies showing it can reduce myocardial dysfunction in isolated rat hearts subjected to ischemia and reperfusion.
  • Selective TRPM4 inhibitor with an IC50 of 17 μM.
  • Shows no inhibitory activity on TRPM5, TRPC6, and CFTR.
  • Useful for research into ischemia-reperfusion injury.
  • Can reduce myocardial dysfunction in isolated rat hearts.
  • Leads to significant improvement in heart function.
  • Reduces infarct size in affected hearts.