Learn More
Alkali Scientific 9 inch Pasteur pipettes, glass, case of 1000
SDP
Catalog No. 5000662059
Encompass_Preferred
9 inch glass Pasteur pipettes designed for accurate liquid transfer in laboratory environments. Made from high quality glass for durability and chemical resistance, these pipettes feature a tapered tip that allows controlled dispensing of small liquid volumes. The transparent glass construction provides clear visibility during sample handling and solution preparation. Suitable for use in research laboratories, clinical environments, and educational institutions, these pipettes support a wide range of routine laboratory procedures. The 9 inch length allows comfortable handling and improved reach when working with containers, test tubes, and culture vessels. Supplied in a case of 1000 to ensure laboratories have a reliable supply for regular use.
Catalog No. 50-006-62059 Supplier Alkali Scientific Supplier No. LD450900
9 inch glass Pasteur pipettes designed for accurate liquid transfer in laboratory environments. Made from high quality glass for durability and chemical resistance, these pipettes feature a tapered tip that allows controlled dispensing of small liquid volumes. The transparent glass construction provides clear visibility during sample handling and solution preparation. Suitable for use in research laboratories, clinical environments, and educational institutions, these pipettes support a wide range of routine laboratory procedures. The 9 inch length allows comfortable handling and improved reach when working with containers, test tubes, and culture vessels. Supplied in a case of 1000 to ensure laboratories have a reliable supply for regular use.