Learn More
Medchemexpress LLC 9-Ing-41 (Elraglusib) | 1034895-42-5 | 404.35 | 10 MG
SDP
Catalog No. 5000436468
Encompass_Preferred
9-ING-41 (Elraglusib) is a maleimide-based ATP-competitive and selective glycogen synthase kinase-3β (GSK-3β) inhibitor with an IC50 of 0.71 μM. This compound leads to significant cell cycle arrest, autophagy, and apoptosis in cancer cells. It demonstrates anticancer activity and has potential for enhancing the antitumor effects of chemotherapeutic agents.
  • Maleimide-based ATP-competitive and selective glycogen synthase kinase-3β (GSK-3β) inhibitor
  • IC50 of 0.71 μM against GSK-3β
  • Significantly induces cell cycle arrest in cancer cells
  • Promotes autophagy in cancer cells
  • Leads to apoptosis in cancer cells
  • Exhibits anticancer activity
  • Potential to enhance antitumor effects of chemotherapeutic agents
Catalog No. 50-004-36468 Supplier Medchemexpress LLC Supplier No. HY11391410MG
9-ING-41 (Elraglusib) is a maleimide-based ATP-competitive and selective glycogen synthase kinase-3β (GSK-3β) inhibitor with an IC50 of 0.71 μM. This compound leads to significant cell cycle arrest, autophagy, and apoptosis in cancer cells. It demonstrates anticancer activity and has potential for enhancing the antitumor effects of chemotherapeutic agents.
  • Maleimide-based ATP-competitive and selective glycogen synthase kinase-3β (GSK-3β) inhibitor
  • IC50 of 0.71 μM against GSK-3β
  • Significantly induces cell cycle arrest in cancer cells
  • Promotes autophagy in cancer cells
  • Leads to apoptosis in cancer cells
  • Exhibits anticancer activity
  • Potential to enhance antitumor effects of chemotherapeutic agents