Learn More
Medchemexpress LLC 9-ING-41 (Elraglusib) | 1034895-42-5 | 99.1% | 404.35 | 1 ML
SDP
Catalog No. 5000436581
Encompass_Preferred
9-ING-41 (Elraglusib) is a maleimide-based ATP-competitive and selective glycogen synthase kinase-3β (GSK-3β) inhibitor. With an IC50 of 0.71 μM, it leads to cell cycle arrest, autophagy, and apoptosis in cancer cells. This compound exhibits significant anticancer activity and has the potential to enhance the antitumor effects of various chemotherapeutic agents.
  • Maleimide-based ATP-competitive and selective GSK-3β inhibitor
  • Induces cell cycle arrest
  • Promotes autophagy
  • Induces apoptosis in cancer cells
  • Exhibits anticancer activity
  • Enhances antitumor effects of chemotherapeutic agents
Catalog No. 50-004-36581 Supplier Medchemexpress LLC Supplier No. HY11391410MM/1ML
9-ING-41 (Elraglusib) is a maleimide-based ATP-competitive and selective glycogen synthase kinase-3β (GSK-3β) inhibitor. With an IC50 of 0.71 μM, it leads to cell cycle arrest, autophagy, and apoptosis in cancer cells. This compound exhibits significant anticancer activity and has the potential to enhance the antitumor effects of various chemotherapeutic agents.
  • Maleimide-based ATP-competitive and selective GSK-3β inhibitor
  • Induces cell cycle arrest
  • Promotes autophagy
  • Induces apoptosis in cancer cells
  • Exhibits anticancer activity
  • Enhances antitumor effects of chemotherapeutic agents