Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Julolidinecarboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. J00081G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
J00081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. J00081G Supplier TCI America Supplier No. J00081G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 33985-71-6
Molecular Formula C13H15NO
Molecular Weight (g/mol) 201.269
MDL Number MFCD00151555
InChI Key XIIVBURSIWWDEO-UHFFFAOYSA-N
Synonym 2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizine-9-carboxaldehyde
PubChem CID 98700
SMILES C1CC2=CC(=CC3=C2N(C1)CCC3)C=O

Specifications

Melting Point 83°C
Color Yellow
Quantity 1 g
Formula Weight 201.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Julolidinecarboxaldehyde
RTECSNumber DK1480000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.