Learn More
Cayman Chemical 9-keto Fluprostenol 10mg
SDP
Catalog No. 502968238
Encompass_Preferred
9-keto Fluprostenol is an analog of PGE2 with structural modifications intended to give it a prolonged half-life and greater potency. Fluprostenol is a well-studied, potent analog of PGF2α and acts primarily through the FP receptor.{1182} Oxidation at C-9 of fluprostenol yields 9-keto fluprostenol. It is anticipated that this analog will have strong affinity for EP receptors and act as a PGE2 agonist.
Catalog No. 50-296-8238 Supplier Cayman Chemical Supplier No. 1678110 MG
9-keto Fluprostenol is an analog of PGE2 with structural modifications intended to give it a prolonged half-life and greater potency. Fluprostenol is a well-studied, potent analog of PGF2α and acts primarily through the FP receptor.{1182} Oxidation at C-9 of fluprostenol yields 9-keto fluprostenol. It is anticipated that this analog will have strong affinity for EP receptors and act as a PGE2 agonist.