Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 9-Mercapto-1-nonanol

Catalog No. 6987681G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-876-81G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-876-81G Supplier Sigma Aldrich Supplier No. 6987681G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

9-Mercapto-1-nonanol (9-MNL) is an alkanethiol that forms a self-assembled monolayer (SAM) on a variety of substrates. It facilitates the immobilization of the surface atoms.9-MNL can be used in the surface modification of gold electrodes for the fabrication of biosensors for biomedical applications.

Specifications

CAS 131215-92-4
Density 0.939 g/mL (at 25°C)
Molecular Formula C9H20OS
Refractive Index n20/D 1.483
Linear Formula C9H20OS
Quantity 1 g
MDL Number MFCD11044866
Synonym 9-Mercapto-nonan-1-ol
Molecular Weight (g/mol) 176.32
Percent Purity 96%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.