Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 9-Mesityl-10-Methylacridinium Tetrafluoroborate

Catalog No. 794171250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-417-1250MG 1 g
79-417-11G 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 79-417-1250MG Supplier Sigma Aldrich Supplier No. 794171250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Catalyst has been reported by Nicewicz and coworkers to mediate myriad transformations through Photoredox Catalysis. The following applications include the anti-Markovnikov hydroamination of alkenes and addition of carboxylic acids to alkenes and the hydrotrifluoromethylation of styrenes using the Langlois reagent (Aldrich 743232).Direct Catalytic Anti-Markovnikov Addition of Carboxylic Acids to Alkenes

Specifications

CAS 1442433-71-7
Molecular Formula C23H22BF4N
Linear Formula C23H22BF4N
Quantity 1 g
Synonym 9-Mesityl-10-methylacridinium tetrafluoroborate; 10-Methyl-9-(2,4,6-trimethylphenyl)acridinium tetrafluoroborate
Molecular Weight (g/mol) 399.23 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.