Learn More
Medchemexpress LLC HY-112642 10mg Medchemexpress, 9-Methoxycanthin-6-one CAS:74991-91-6 Purity:>98%
SDP
Catalog No. 501969127
Encompass_Preferred
Medchemexpress, HY-112642 10mg 9-Methoxycanthin-6-one CAS:74991-91-6 9-Methoxycanthin-6-one, a canthin-6-one alkaloid, is present in intact plant parts and in callus tissues of different explants. 9-Methoxycanthin-6-one shows anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9127 Supplier Medchemexpress LLC Supplier No. HY11264210MG
Medchemexpress, HY-112642 10mg 9-Methoxycanthin-6-one CAS:74991-91-6 9-Methoxycanthin-6-one, a canthin-6-one alkaloid, is present in intact plant parts and in callus tissues of different explants. 9-Methoxycanthin-6-one shows anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.