Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 9-Methyl-9H-Fluorene-9-Carbonyl Chloride

Catalog No. 7450225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-502-25G 5 g
74-502-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 74-502-25G Supplier Sigma Aldrich Supplier No. 7450225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

COGen is a simple alternative for carbon monoxide generation in the application of palladium-catalyzed carbonylation reactions. This reagent has proved to be applicable in a wide range of applications and is compatible with several building blocks in the formation of ketones, amides, esters, etc.For more information please visit: Technology Spotlight, Professor Skrystrup PPP

Specifications

CAS 82102-37-2
Molecular Formula C15H11ClO
Linear Formula C15H11ClO
MDL Number MFCD18839248
Quantity 5 g
Synonym COgen; 9-Methylfluorene-9-carbonyl chloride
Molecular Weight (g/mol) 242.7 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.