Learn More
TARGETMOL CHEMICALS INC 9-Methyl salvianolate B 5MG
SDP
Catalog No. 502879960
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 9'-Methyl lithospermate B is a phenolic compound extracted from Radix Salvia miltiorrhizae. Purity 97.25%
Catalog No. 50-287-9960 Supplier TARGETMOL CHEMICALS INC Supplier No. T49615MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 9'-Methyl lithospermate B is a phenolic compound extracted from Radix Salvia miltiorrhizae. Purity 97.25%