Learn More
AdipoGen 9-Methylguanine
SDP
Catalog No. 502560526
Encompass_Preferred
Chemical. CAS 5502-78-3. Formula C6H7N5O. MW 165.15. Synthetic Nucleic acid base useful for synthesis and complex building. Catalyzes the conversion of 7,8-dihydroneopterin to 6- hydroxymethyl-7,8-dihydropterin.
Catalog No. 50-256-0526 Supplier AdipoGen Supplier No. CDXM0095M100
Chemical. CAS 5502-78-3. Formula C6H7N5O. MW 165.15. Synthetic Nucleic acid base useful for synthesis and complex building. Catalyzes the conversion of 7,8-dihydroneopterin to 6- hydroxymethyl-7,8-dihydropterin.