Learn More
Waters Corp Waters Cap, 9 mm, Black polypropylene, PTFE/Silicone Liner
Catalog No. 50596975 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 9 mm solid black cap features a PTFE/silicone liner, designed for use with 12 x 32 mm vials for sample storage.
  • Made from high-quality polypropylene material.
  • Designed to securely seal both liquid and solid samples.
  • Permanently attached PTFE silicone liner ensures a tight seal.
  • Can be safely stored within a temperature range of -40°C to 100°C.
  • Compatible with most standard autosamplers.
Catalog No. 50-596-975 Supplier Waters Corp Supplier No. 186007187
This 9 mm solid black cap features a PTFE/silicone liner, designed for use with 12 x 32 mm vials for sample storage.
  • Made from high-quality polypropylene material.
  • Designed to securely seal both liquid and solid samples.
  • Permanently attached PTFE silicone liner ensures a tight seal.
  • Can be safely stored within a temperature range of -40°C to 100°C.
  • Compatible with most standard autosamplers.