Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Nitroanthracene, MP Biomedicals

Catalog No. ICN214441 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN214441 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN214441 Supplier MP Biomedicals, Inc Supplier No. 0521444105

Chemical Identifiers

CAS 602-60-8
Molecular Formula C14H9NO2
Molecular Weight (g/mol) 223.231
InChI Key LSIKFJXEYJIZNB-UHFFFAOYSA-N
Synonym anthracene, 9-nitro, 5-nitroanthracene, unii-jz1tb3n57p, 9-nitro-anthracene, ccris 4679, jz1tb3n57p, 9-nitro anthracene, pubchem15119, acmc-20ab5c, 9-nitroanthracene
PubChem CID 11767
ChEBI CHEBI:82501
IUPAC Name 9-nitroanthracene
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2[N+](=O)[O-]

Specifications

Quantity 5 g
Formula Weight 223.23
Chemical Name or Material 9-Nitroanthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.