Learn More
Cayman Chemical 9-Anthryldiazomethane, Purity: ≥85%, CAS: 10401-59-9, 10 mg
SDP
Catalog No. 502935355
Encompass_Preferred
A fluorescent probe; has been used for the detection of fatty acids, arachidonic acid metabolites, prostaglandins, leukotrienes, and thromboxanes; ex/em = 365/412 nm in 80:20 solutions of acetonitrile:water and methanol:water; has been used to detect amino acids via HPLC with ex/em = 255/412 nm, respectively; has been used as a synthetic intermediate in the synthesis of anthracene-containing polymers
Catalog No. 50-293-5355 Supplier Cayman Chemical Supplier No. 2680610 MG
A fluorescent probe; has been used for the detection of fatty acids, arachidonic acid metabolites, prostaglandins, leukotrienes, and thromboxanes; ex/em = 365/412 nm in 80:20 solutions of acetonitrile:water and methanol:water; has been used to detect amino acids via HPLC with ex/em = 255/412 nm, respectively; has been used as a synthetic intermediate in the synthesis of anthracene-containing polymers