Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9-octadecenoic acid (9z)-, 2,3-dihydroxypropyl-1,1,2,3,3-d5 ester | 565183-24-6 | 99.9% | 10 MG
SDP

Supplier:  Medchemexpress LLC HY128754S10MG

Encompass_Preferred

Monoolein-d5 is a deuterium labeled monoolein with high purity, making it suitable for research applications requiring isotopic labeling. Its structure has been confirmed by 1H NMR and mass spectrometry.

  • Purity of 99.9% (HPLC)
  • Isotopic enrichment of 99.9%
  • Structure confirmed by 1H NMR and MS
  • Store at 4°C, sealed, away from moisture and light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-42925


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.