Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9-Octadecenoic acid (Z)-, 1,2,3-propanetriyl-1,1,2,3,3-d5 ester | 60892-83-3 | MFCD18782961 | 97.0% | 890.46 | C57H99D5O6 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN1981S110MG

Encompass_Preferred

Triolein-d5 is the deuterium-labeled form of triolein, provided as a high-purity stable isotope standard for research and analytical applications such as LC-MS and GC-MS quantification. It is a symmetrical triacylglycerol reported to reduce MMP-1 upregulation and to possess antioxidant and anti-inflammatory activity.

  • Deuterium-labeled stable isotope standard for analytical use
  • Chemical formula C57H99D5O6 and molecular weight 890.46
  • Purity 97.0%
  • Supplied as a 10 mg vial
  • Soluble in DMSO (100 mg/mL); sonication recommended
  • Store pure material at -20°C (3 years) or 4°C (2 years); in solvent store at -80°C (6 months) or -20°C (1 month)

Catalog No. 50-002-95486


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.