Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9-octadecynoic acid | 506-24-1 | MFCD00014386 | 99.9% | 280.45 g/mol | C18H32O2 | 1 ML
SDP

Catalog No. 5000282101
Encompass_Preferred

9-Octadecynoic acid is a fatty-acid-derived small molecule reported to bind DNA (Kd ≈ 1.8 mM) and act as an agonist of peroxisome proliferator-activated receptor γ (PPARγ). It is supplied as a 10 mM stock solution in DMSO (1 mL) for research use.

  • DNA-binding agent with reported Kd ≈ 1.8 mM.
  • Agonist of PPARγ, useful for receptor modulation studies.
  • Supplied as a 10 mM DMSO stock solution (1 mL) for convenient dosing.
  • High purity (99.9%) suitable for biochemical assays.
  • Molecular weight 280.45 g/mol; formula C18H32O2.
  • Store in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-82101 Supplier Medchemexpress LLC Supplier No. HYW34199710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

9-Octadecynoic acid is a fatty-acid-derived small molecule reported to bind DNA (Kd ≈ 1.8 mM) and act as an agonist of peroxisome proliferator-activated receptor γ (PPARγ). It is supplied as a 10 mM stock solution in DMSO (1 mL) for research use.

  • DNA-binding agent with reported Kd ≈ 1.8 mM.
  • Agonist of PPARγ, useful for receptor modulation studies.
  • Supplied as a 10 mM DMSO stock solution (1 mL) for convenient dosing.
  • High purity (99.9%) suitable for biochemical assays.
  • Molecular weight 280.45 g/mol; formula C18H32O2.
  • Store in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.