Learn More
Medchemexpress LLC 9-octadecynoic acid | 506-24-1 | MFCD00014386 | 99.9% | 280.45 g/mol | C18H32O2 | 50 MG
SDP
Catalog No. 5000282548
Encompass_Preferred
9-Octadecynoic acid is an acetylenic fatty acid used in biochemical research as a DNA-binding agent and as an agonist of peroxisome proliferator-activated receptor γ (PPARγ). It is characterized by an 18-carbon backbone and is supplied for in vitro studies that require high purity and defined physicochemical properties.
  • Acts as a DNA binding agent with reported Kd ≈ 1.8 mM.
  • Functions as a PPARγ agonist in biological assays.
  • Chemical formula C18H32O2; molecular weight 280.45 g/mol.
  • High purity (99.9%) suitable for research applications.
  • Available as a solid and as solution formulations (for example, 10 mM in DMSO).
Catalog No. 50-002-82548 Supplier Medchemexpress LLC Supplier No. HYW34199750MG
9-Octadecynoic acid is an acetylenic fatty acid used in biochemical research as a DNA-binding agent and as an agonist of peroxisome proliferator-activated receptor γ (PPARγ). It is characterized by an 18-carbon backbone and is supplied for in vitro studies that require high purity and defined physicochemical properties.
  • Acts as a DNA binding agent with reported Kd ≈ 1.8 mM.
  • Functions as a PPARγ agonist in biological assays.
  • Chemical formula C18H32O2; molecular weight 280.45 g/mol.
  • High purity (99.9%) suitable for research applications.
  • Available as a solid and as solution formulations (for example, 10 mM in DMSO).