Learn More
Medchemexpress LLC 9-oxo-9H-indeno [1,2-b]pyrazine-2,3-dicarbonitrile | 924296-17-3 | 99.0% | 287.28 g·mol⁻¹ | C16H9N5O | 10 MG
SDP
Catalog No. 5000297943
Encompass_Preferred
DUB-IN-3 is a small-molecule deubiquitinase inhibitor reported to selectively inhibit USP8 and used in biochemical and cell-based research. Supplied as a light yellow solid with reported purity ≥99% and molecular weight 287.28 g·mol⁻¹, it is intended for research use only.
  • Potent USP8 inhibitor with reported IC50 ≈ 0.56 μM.
  • High purity suitable for research applications.
  • Available as a 10 mg solid and in larger pack sizes or solution forms.
  • Light yellow to yellow solid with defined storage conditions for powder and solution.
  • Documented CAS number and molecular formula for unambiguous identification.
Catalog No. 50-002-97943 Supplier Medchemexpress LLC Supplier No. HY5073710MG
DUB-IN-3 is a small-molecule deubiquitinase inhibitor reported to selectively inhibit USP8 and used in biochemical and cell-based research. Supplied as a light yellow solid with reported purity ≥99% and molecular weight 287.28 g·mol⁻¹, it is intended for research use only.
  • Potent USP8 inhibitor with reported IC50 ≈ 0.56 μM.
  • High purity suitable for research applications.
  • Available as a 10 mg solid and in larger pack sizes or solution forms.
  • Light yellow to yellow solid with defined storage conditions for powder and solution.
  • Documented CAS number and molecular formula for unambiguous identification.